MilliporeSigma
  • Olfactory receptor cells respond to odors in a tissue and semiconductor hybrid neuron chip.
Olfactory receptor cells respond to odors in a tissue and semiconductor hybrid neuron chip.
Biosensors & bioelectronics (2010-10-15)
Qingjun Liu, Weiwei Ye, Ning Hu, Hua Cai, Hui Yu, Ping Wang
ABSTRACT
Olfactory systems of human beings and animals have the abilities to sense and distinguish varieties of odors. In this study, a bioelectronic nose was constructed by fixing biological tissues onto the surface of light-addressable potentiometric sensor (LAPS) to mimic human olfaction and realize odor differentiation. The odorant induced potentials on tissue-semiconductor interface was analyzed by sensory transduction theory and sheet conductor model. The extracellular potentials of the receptor cells in the olfactory epithelium were detected by LAPS. Being stimulated by different odorants, such as acetic acid and butanedione, olfactory epithelium activities were analyzed on basis of local field potentials and presented different firing modes. The signals fired in different odorants could be distinguished into different clusters by principal component analysis (PCA). Therefore, with cellular populations well preserved, the epithelium tissue and LAPS hybrid system will be a promising neuron chip of olfactory biosensors for odor detecting.
Sigma-Aldrich
1,3-Butadiene diepoxide, 97%