MilliporeSigma
  • "Nanoarmoured" droplets of different shapes formed by interfacial self-assembly and crosslinking of metal nanoparticles.
"Nanoarmoured" droplets of different shapes formed by interfacial self-assembly and crosslinking of metal nanoparticles.
Nanoscale (2010-10-13)
Bartlomiej Kowalczyk, István Lagzi, Bartosz A Grzybowski
ABSTRACT
Films comprising metal nanoparticles are assembled on the surfaces of liquid droplets of different shapes and macroscopic dimensions. These films are reinforced by dithiol crosslinks and are mechanically rugged yet permeable to the diffusion of small molecules.
Sigma-Aldrich
Bromothymol Blue, ACS reagent, Dye content 95 %