MilliporeSigma
  • Sustained pain freedom and no adverse events as an endpoint in clinical trials of acute migraine treatments: application to patient-level data from a trial of the CGRP receptor antagonist, telcagepant, and zolmitriptan.
Sustained pain freedom and no adverse events as an endpoint in clinical trials of acute migraine treatments: application to patient-level data from a trial of the CGRP receptor antagonist, telcagepant, and zolmitriptan.
Cephalalgia : an international journal of headache (2010-10-13)
David W Dodick, James Kost, Christopher Assaid, Christopher Lines, Tony W Ho
ABSTRACT
Endpoints used to evaluate the efficacy of acute anti-migraine drugs do not measure the tolerability. Sustained pain-free response with no adverse events has been recommended as a composite endpoint which measures the efficacy and tolerability attributes that patients desire. The aim of this study was to evaluate new composite efficacy-plus-tolerability endpoints based on a post-hoc analysis of patient-level data from a previous randomized, placebo-controlled trial of the calcitonin gene-related peptide (CGRP) receptor antagonist, telcagepant, and zolmitriptan in the acute treatment of migraine. Endpoints were 2-24-hour sustained pain freedom and no adverse events from 0-24 hours (SPF24NAE), 2-24 hour sustained pain relief and no adverse events from 0-24 hours (SPR24NAE), pain freedom at 2 hours and no adverse events from 0-24 hours (PF2NAE), and pain relief at 2 hours and no adverse events from 0-24 hours (PR2NAE). Compared with placebo, both telcagepant 300 mg and 150 mg achieved nominal superiority (p values <.05) for SPF24NAE, SPR24NAE, PF2NAE and PR2NAE. Zolmitriptan 5 mg showed nominal superiority versus placebo for SPF24NAE, SPR24NAE and PF2NAE, but not PR2NAE. Telcagepant 300 mg showed nominal superiority versus zolmitriptan for SPF24NAE, SPR24NA and PR2NAE. Composite efficacy-plus-tolerability endpoints may be useful for facilitating comparisons between treatments.
Sigma-Aldrich
Zolmitriptan, ≥98% (HPLC)