MilliporeSigma
  • InCl3-catalyzed allylic Friedel-Crafts reactions toward the stereocontrolled synthesis of 1,2,3,4-tetrahydroquinolines.
InCl3-catalyzed allylic Friedel-Crafts reactions toward the stereocontrolled synthesis of 1,2,3,4-tetrahydroquinolines.
Organic letters (2010-10-01)
David S Giera, Christoph Schneider
ABSTRACT
Allyl chlorides tethered to an N-aryl moiety readily undergo InCl(3)-catalyzed Friedel-Crafts reactions to furnish highly enantiomerically enriched 1,2,3,4-tetrahydroquinolines with good yields and excellent diastereoselectivity.
Sigma-Aldrich
Indium(III) chloride, anhydrous, powder, ≥99.999% trace metals basis
Sigma-Aldrich
Indium(III) chloride, 99.999% trace metals basis
Sigma-Aldrich
Indium(III) chloride, 98%