MilliporeSigma
Synthesis of enantiopure 3-substituted morpholines.
The Journal of organic chemistry (2010-10-01)
Jan Bornholdt, Jakob Felding, Jesper Langgaard Kristensen
ABSTRACT
Enantiopure 3-substituted morpholines were assembled through ring-opening of a N-2-benzothiazolesulfonyl (Bts) activated aziridine with organocuprates followed by a ring annulation reaction with a vinylsulfonium salt under microwave conditions. Deprotection of the N-Bts group proceeds under very mild conditions with 2-mercaptoacetic acid and LiOH at rt.
Sigma-Aldrich
Morpholine, ReagentPlus®, ≥99%
Sigma-Aldrich
Morpholine, ACS reagent, ≥99.0%
Sigma-Aldrich
Morpholine, purified by redistillation, ≥99.5%
Supelco
Morpholine, analytical standard