MilliporeSigma
  • Synthesis of amidomethyltrifluoroborates and their use in cross-coupling reactions.
Synthesis of amidomethyltrifluoroborates and their use in cross-coupling reactions.
Organic letters (2010-10-01)
Gary A Molander, Marie-Aude Hiebel
ABSTRACT
Amidomethyltrifluoroborates were successfully synthesized in a one-pot fashion and used in cross-coupling reactions with a wide variety of aryl and heteroaryl chlorides.
Sigma-Aldrich
Tetrafluoroboric acid solution, 48 wt. % in H2O