MilliporeSigma
  • Cytotoxicity on L1210 leukemic cells of beta-amanitin-concanavalin A and phallacidin-concanavalin A conjugates.
Cytotoxicity on L1210 leukemic cells of beta-amanitin-concanavalin A and phallacidin-concanavalin A conjugates.
Toxicon : official journal of the International Society on Toxinology (1990-01-01)
Z Zhelev, M Ilarionova, A Zheleva, C Alexiev, K Halacheva, D Todorov
ABSTRACT
The conjugates beta-amanitin-concanavalin A and phallacidin concanavalin A were tested for direct cytotoxicity on L1210 lymphocytic leukemia cells by a combined in vitro-in vivo bioassay. Both conjugates exerted strong direct cytotoxicity on the tumour cells.
Sigma-Aldrich
β-Amanitin from Amanita phalloides, ≥85% (HPLC)