MilliporeSigma
  • Coated cross-species antibodies by mannosamine-biotin adduct confer protection against snake venom without eliciting humoral immune response.
Coated cross-species antibodies by mannosamine-biotin adduct confer protection against snake venom without eliciting humoral immune response.
Vaccine (2010-09-30)
Tal Gefen, Jacob Pitcovski, Jacob Vaya, Soliman Khatib, Simi Krispel, E Dan Heller, Elena Gaberman, Raphael Gorodetsky, Elina Aizenshtein
ABSTRACT
Passive immunization with cross-species antibodies triggers the patient's immune response, thereby preventing repeated treatment. Mannosamine-biotin adduct (MBA) has been described as a masking agent for immunogenic reduction and here, the immunogenicity and biological activity of MBA-coated horse anti-viper venom (hsIgG) were compared to those of uncoated or PEGylated hsIgG. In in vitro tests, hsIgG binding was not affected by MBA conjugation. The immune response to hsIgG-MBA was about 8-fold and 32-fold lower than to PEG-coated and uncoated hsIgG, respectively. In vivo, hsIgG-MBA showed efficient venom-neutralization activity. We thus demonstrate the feasibility of using MBA as a masking agent for passive immunization with cross-species antibodies.
Sigma-Aldrich
D-Mannosamine hydrochloride, ≥98% (HPLC)