MilliporeSigma
  • Pharmacokinetics and metabolism of natural methylxanthines in animal and man.
Pharmacokinetics and metabolism of natural methylxanthines in animal and man.
Handbook of experimental pharmacology (2010-09-23)
Maurice J Arnaud
ABSTRACT
Caffeine, theophylline, theobromine, and paraxanthine administered to animals and humans distribute in all body fluids and cross all biological membranes. They do not accumulate in organs or tissues and are extensively metabolized by the liver, with less than 2% of caffeine administered excreted unchanged in human urine. Dose-independent and dose-dependent pharmacokinetics of caffeine and other dimethylxanthines may be observed and explained by saturation of metabolic pathways and impaired elimination due to the immaturity of hepatic enzyme and liver diseases. While gender and menstrual cycle have little effect on their elimination, decreased clearance is seen in women using oral contraceptives and during pregnancy. Obesity, physical exercise, diseases, and particularly smoking and the interactions of drugs affect their elimination owing to either stimulation or inhibition of CYP1A2. Their metabolic pathways exhibit important quantitative and qualitative differences in animal species and man. Chronic ingestion or restriction of caffeine intake in man has a small effect on their disposition, but dietary constituents, including broccoli and herbal tea, as well as alcohol were shown to modify their plasma pharmacokinetics. Using molar ratios of metabolites in plasma and/or urine, phenotyping of various enzyme activities, such as cytochrome monooxygenases, N-acetylation, 8-hydroxylation, and xanthine oxidase, has become a valuable tool to identify polymorphisms and to understand individual variations and potential associations with health risks in epidemiological surveys.
Product Description
Supelco
Paraxanthine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
1,7-Dimethylxanthine, ~98%, solid