MilliporeSigma
  • GMP-grade pneumococcal whole-cell vaccine injected subcutaneously protects mice from nasopharyngeal colonization and fatal aspiration-sepsis.
GMP-grade pneumococcal whole-cell vaccine injected subcutaneously protects mice from nasopharyngeal colonization and fatal aspiration-sepsis.
Vaccine (2010-09-23)
Ying-Jie Lu, Luciana Leite, Viviane Maimoni Gonçalves, Waldely de Oliveira Dias, Celia Liberman, Fernando Fratelli, Mark Alderson, Andrea Tate, Jean-François Maisonneuve, George Robertson, Rita Graca, Sabina Sayeed, Claudette M Thompson, Porter Anderson, Richard Malley
ABSTRACT
Mucosal immunization with a killed whole-cell pneumococcal vaccine, given with enterotoxin-related adjuvants, has been shown to confer multi-serotype protection against colonization of the nasopharynx and middle ear in mice. However, because novel mucosal immunization strategies may be difficult to implement, here we evaluated subcutaneous injection. Strain RM200 was engineered to be capsule-negative, autolysin-negative, and to express a non-toxic mutant pneumolysoid. Liter-scale and 60-l Good Manufacturing Practice (GMP) cultures were grown in bovine-free soy-based medium, killed with chloroform or beta-propiolactone, and injected into C57Bl/6 mice without or with aluminum adjuvant. The adjuvant Al(OH)(3) strongly increased responses, particularly if pre-treated with phosphate. Protection was found in several tested model infections: nasal colonization with a serotype 6B strain and fatal aspiration-sepsis with strains of serotype 3 and 5. Protection against colonization was mechanistically dependent on the presence of CD4+ T cells at the time of challenge; in contrast, in the type 3 aspiration-sepsis model, CD4+ T cells were not required for protection at the time of challenge, suggesting that antibody alone was sufficient to protect against death in this model. Rabbits receiving sequential intramuscular injections in a pilot toxicity study displayed local reactogenicity at injection sites but no clinical signs. The rabbit antiserum thus produced was active in an in vitro phagocytic killing assay and passively protected mice in the type 3 aspiration-sepsis model. Approval is being sought for human trials of this vaccine.
Sigma-Aldrich
β-Propiolactone, Grade II, ≥90%