MilliporeSigma
  • Synthesis of heterocycles via Pd-ligand controlled cyclization of 2-chloro-N-(2-vinyl)aniline: preparation of carbazoles, indoles, dibenzazepines, and acridines.
Synthesis of heterocycles via Pd-ligand controlled cyclization of 2-chloro-N-(2-vinyl)aniline: preparation of carbazoles, indoles, dibenzazepines, and acridines.
Journal of the American Chemical Society (2010-09-23)
Dmitry Tsvelikhovsky, Stephen L Buchwald
ABSTRACT
The Pd-catalyzed condensation of 2-bromostyrene and 2-chloroaniline derivatives yields stable diphenylamine intermediates, which are selectively converted to five-, six-, or seven-membered heteroaromatics (indoles, carbazoles, acridines, and dibenzazepines). The selectivity of these intramolecular transformations is uniquely ligand-controlled and offers efficient routes to four important classes of heterocycles from a common precursor.
Sigma-Aldrich
2-Chloroaniline, ≥99.5% (GC)
Sigma-Aldrich
2-Chloroaniline, technical, ≥98.0% (GC)