MilliporeSigma
  • Molecular biology, phytochemistry and bioactivity of three endemic Aloe species from Mauritius and Réunion Islands.
Molecular biology, phytochemistry and bioactivity of three endemic Aloe species from Mauritius and Réunion Islands.
Phytochemical analysis : PCA (2010-09-08)
Mala Ranghoo-Sanmukhiya, Joyce Govinden-Soulange, Christophe Lavergne, Shannoo Khoyratty, Denis Da Silva, Michel Frederich, Hippolyte Kodja
ABSTRACT
Aloe tormentorii, A. purpurea and A. macra are used as multipurpose folk medicines in Réunion and Mauritius Islands and are mistaken for the introduced Aloe vera. To compare the phytochemical, antimicrobial and DNA profiles of Aloe endemic to Mauritius and Réunion with the profiles of A. vera. Methodology - Leaf extracts of these Aloe species were analysed using standard phytochemical screening techniques, TLC and by HPLC. These extracts were also assayed for antimicrobial activity using microdilution techniques. Genetic diversity was studied using RAPD markers. Phytochemical and antimicrobial assays and RAPD analysis showed that Mascarene Aloe species were very different from A. vera. This study is the first report highlighting the differences between Aloe sp.p from Mascarene and Aloe vera at the metabolic and genomic level.
Product Description
Isoorientin, primary reference standard
Sigma-Aldrich
Isoorientin, ≥98% (HPLC)