MilliporeSigma
Purinergic inhibition of ENaC produces aldosterone escape.
Journal of the American Society of Nephrology : JASN (2010-09-04)
James D Stockand, Elena Mironova, Vladislav Bugaj, Timo Rieg, Paul A Insel, Volker Vallon, Janos Peti-Peterdi, Oleh Pochynyuk
ABSTRACT
The mechanisms underlying "aldosterone escape," which refers to the excretion of sodium (Na(+)) during high Na(+) intake despite inappropriately increased levels of mineralocorticoids, are incompletely understood. Because local purinergic tone in the aldosterone-sensitive distal nephron downregulates epithelial Na(+) channel (ENaC) activity, we tested whether this mechanism mediates aldosterone escape. Here, urinary ATP concentration increased with dietary Na(+) intake in mice. Physiologic concentrations of ATP decreased ENaC activity in a dosage-dependent manner. P2Y(2)(-/-) mice, which lack the purinergic receptor, had significantly less increased Na(+) excretion than wild-type mice in response to high-Na(+) intake. Exogenous deoxycorticosterone acetate and deletion of the P2Y(2) receptor each modestly increased the resistance of ENaC to changes in Na(+) intake; together, they markedly increased resistance. Under the latter condition, ENaC could not respond to changes in Na(+) intake. In contrast, as a result of aldosterone escape, wild-type mice had increased Na(+) excretion in response to high-Na(+) intake regardless of the presence of high deoxycorticosterone acetate. These data suggest that control of ENaC by purinergic signaling is necessary for aldosterone escape.
Product Description
Desoxycortone acetate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Deoxycorticosterone acetate