MilliporeSigma
  • One-pot microwave-assisted protocol for the synthesis of substituted 2-amino-1H-imidazoles.
One-pot microwave-assisted protocol for the synthesis of substituted 2-amino-1H-imidazoles.
Molecular diversity (2010-08-27)
D S Ermolat'ev, B Savaliya, A Shah, E Van der Eycken
ABSTRACT
An efficient microwave-assisted one-pot two-step protocol was developed for the construction of disubstituted 2-amino-1H-imidazoles. This process involves the sequential formation of 2,3-dihydro-2-hydroxyimidazo[1,2-a]pyrimidinium salts from readily available 2-aminopyrimidines and α-bromoketones, followed by cleavage of the pyrimidine ring with hydrazine.
Sigma-Aldrich
2-Aminopyrimidine, 97%