MilliporeSigma
  • Interfacial complexes between a protein and lipophilic ions at an oil-water interface.
Interfacial complexes between a protein and lipophilic ions at an oil-water interface.
Analytical chemistry (2010-08-26)
Rune A Hartvig, Manuel A Méndez, Marco van de Weert, Lene Jorgensen, Jesper Østergaard, Hubert H Girault, Henrik Jensen
ABSTRACT
The interaction between an intact protein and two lipophilic ions at an oil-water interface has been investigated using cyclic voltammetry, impedance based techniques and a newly developed method in which the biphasic oil-water system is analyzed by biphasic electrospray ionization mass spectrometry (BESI-MS), using a dual-channel electrospray emitter. It is found that the protein forms interfacial complexes with the lipophilic ions and that it specifically requires the presence of the oil-water interface to be formed under the experimental conditions. Furthermore, impedance based techniques and BESI-MS with a common ion to polarize the interface indicated that the Galvani potential difference across the oil-water interface significantly influences the interfacial complexation degree. The ability to investigate protein-ligand complexes formed at polarized liquid-liquid interfaces is thus a new analytical method for assessing potential dependent interfacial complexation using a structure elucidating detection principle.
Sigma-Aldrich
Potassium tetraphenylborate, 97%
Sigma-Aldrich
Ammonium tetraphenylborate, 99%
Sigma-Aldrich
Sodium tetraphenylborate, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium tetraphenylborate, puriss. p.a., ACS reagent, ≥99.5% (NT)
Supelco
Potassium tetrakis(4-chlorophenyl)borate, Selectophore, ≥98.0%