MilliporeSigma
  • Diversity-oriented, one-pot, multi-component synthesis of substituted uracil derivatives.
Diversity-oriented, one-pot, multi-component synthesis of substituted uracil derivatives.
Molecular diversity (2010-08-21)
Yogesh Y Pedgaonkar, Mariam S Degani, Radhakrishnan P Iyer
ABSTRACT
With the emergence of high throughput screening of bioactive molecules, there is constant need for the development of new strategies for diversity-oriented synthesis. We describe here a novel one-pot multicomponent reaction for the synthesis of uracil derivatives using easily available starting materials. This new synthetic strategy provides easy access to diverse uracil derivatives in moderate to good yields.
Sigma-Aldrich
Bis(trichloromethyl) carbonate, purum, ≥99.0% (AT)
Sigma-Aldrich
Triphosgene, reagent grade, 98%
Sigma-Aldrich
Cyanoacetamide, 99%