MilliporeSigma
  • Protective effect of trapidil and l-arginine against renal and hepatic toxicity induced by cyclosporine in rats.
Protective effect of trapidil and l-arginine against renal and hepatic toxicity induced by cyclosporine in rats.
Renal failure (2010-08-21)
Neveen A Salem, Emad A Salem, Aref M Maarouf, Mostafa Kamel, Hazem Elgalaly, Mohamed Radwan, Walid A Abd El-Dayem, Mahmoud Eladl
ABSTRACT
Cyclosporine A (CsA) leads to renal and liver injury, production of free radicals and nitric oxide (NO) deficiency. This study investigates the possible protective effects of trapidil and L-arginine against CsA-induced tissue injury. Forty adult male Wistar rats (180 +/- 20 g) were divided into five groups, eight animals in each. The first group served as control, second group served as CsA group, third group served as CsA + trapidil group, fourth group served as CsA + L-arginine group, and fifth group served as CsA + trapidil + L-arginine group. Kidney and liver functions, inflammatory mediators, cytokines, oxidant and antioxidant parameters as well as histopathological studies of renal and liver tissue were assessed in all groups. CsA induced renal and hepatic dysfunction, which was confirmed by laboratory and histopathological examination. Administration of trapidil diminished the renal and liver injury and significantly attenuated the levels of serum creatinine, urea, aspartate aminotransferase (AST), alanine aminotransferase (ALT), interleukin-1beta (IL-1beta), tumor necrosis factor alpha (TNF-alpha), monocyte chemoattractant protein-1 (MCP-1), and oxidative stress, while it significantly elevated the level of serum nitric oxide and the activity of antioxidative stress. L-Arginine gave the same trend as trapidil, but trapidil effect was more pronounced. Coadministration of trapidil + L-arginine significantly ameliorated the toxic effect of CsA, but did not differ significantly from the effect of trapidil alone. Treatment with trapidil or L-arginine diminished the renal and hepatic CsA-induced toxicity. However, the effect of trapidil was more pronounced. Therefore, treatment with trapidil alone may be the most economic and effective as a potential therapeutic agent in CsA injury.
Product Description
Trapidil, European Pharmacopoeia (EP) Reference Standard