MilliporeSigma
  • Enzymatic reactions and synthesis of n-butyl caproate: esterification, transesterification and aminolysis using a recombinant lipase from Geobacillus thermoleovorans CCR11.
Enzymatic reactions and synthesis of n-butyl caproate: esterification, transesterification and aminolysis using a recombinant lipase from Geobacillus thermoleovorans CCR11.
Environmental technology (2010-08-20)
M Guadalupe Sánchez-Otero, Rodolfo Quintana-Castro, Paola C Mora-González, Ofelia Márquez-Molina, Gerardo Valerio-Alfaro, Rosamaría Oliart-Ros
ABSTRACT
The recombinant lipase LipMatCCR11 from the thermophilic strain Geobacillus thermoleovorans CCR11 was applied in the synthesis of n-butyl caproate via transesterification in hexane and xylene. The short chain flavour ester was obtained by alcoholysis from ethyl caproate and n-butyl alcohol and acidolysis from n-butyl butyrate and caproic acid. This enzyme was also used in the condensation reaction from caproic acid and n-butanol. The conversion percentages at equilibrium (Xe) were similar to those obtained with Candida antarctica lipase fraction B (CAL-B) in the same reaction conditions, while lower conversion velocities (k) were attained. LipMatCCR11 reached high conversion percentages in either hexane or xylene as organic media (> 63%); the enzyme was also able to catalyze the aminolysis reaction of ethyl caproate with benzyl amine in hexane obtaining a conversion percentage > 62%.
Sigma-Aldrich
Butyl hexanoate, ≥98%, FG