MilliporeSigma
  • Luminescent cyclometallated iridium(III) bis(quinolylbenzaldehyde) diimine complexes--synthesis, photophysics, electrochemistry, protein cross-linking properties, cytotoxicity and cellular uptake.
Luminescent cyclometallated iridium(III) bis(quinolylbenzaldehyde) diimine complexes--synthesis, photophysics, electrochemistry, protein cross-linking properties, cytotoxicity and cellular uptake.
Dalton transactions (Cambridge, England : 2003) (2010-08-19)
Pui-Kei Lee, Hua-Wei Liu, Shek-Man Yiu, Man-Wai Louie, Kenneth Kam-Wing Lo
ABSTRACT
Four new luminescent cyclometallated iridium(III) bis(quinolylbenzaldehyde) diimine complexes [Ir(qba)(2)(N⁁N)](PF(6)) (Hqba = 4-(2-quinolyl)benzaldehyde, N⁁N = 2,2'-bipyridine, bpy (1); 1,10-phenanthroline, phen (2); 3,4,7,8-tetramethyl-1,10-phenanthroline, Me(4)-phen (3); 4,7-diphenyl-1,10-phenanthroline, Ph(2)-phen (4)) have been synthesised and characterised, and their electronic absorption, emission and electrochemical properties investigated. The X-ray crystal structures of complexes 1 and 2 have been determined. Upon irradiation, complexes 1-4 exhibited intense and long-lived orange-yellow emission in fluid solutions at 298 K and in alcohol glass at 77 K. The emission has been assigned to a triplet intra-ligand ((3)IL) excited state associated with the qba ligand, probably with mixing of some triplet metal-to-ligand charge-transfer ((3)MLCT) (dπ(Ir) →π*(qba)) character. Reductive amination reactions of complexes 1-4 with the protein bovine serum albumin (BSA) afforded the bioconjugates 1-BSA-4-BSA, respectively. Upon photoexcitation, these bioconjugates displayed intense and long-lived (3)MLCT (dπ(Ir) →π*(N⁁C)) emission in aqueous buffer at 298 K. The cross-linked nature of the Ir-BSA bioconjugates has been verified by SDS-PAGE. Additionally, the cytotoxicity of the complexes towards human cervix epithelioid carcinoma (HeLa) cells has been examined by 3-(4,5-dimethyl-2-thiazolyl)-2,5-diphenyltetrazolium bromide (MTT) assays, and the cellular uptake of complex 4 has been investigated by laser-scanning confocal microscopy and flow cytometry.