MilliporeSigma
  • Kinetics independent spectrometric analysis using non-linear calibration modelling and exploitation of concentration gradients generated by a flow-batch system for albumin and total protein determination in blood serum.
Kinetics independent spectrometric analysis using non-linear calibration modelling and exploitation of concentration gradients generated by a flow-batch system for albumin and total protein determination in blood serum.
Talanta (2010-08-04)
Margarida C Souza, Valdomiro L Martins, Luciano F Almeida, Osmundo D Pessoa Neto, Edvaldo N Gaião, Mario Cesar U Araujo
ABSTRACT
An automatic method for kinetics independent spectrometric analysis is proposed in this study. It uses a non-linear calibration model that explores concentration gradients generated by a flow-batch analyser (FBA) for the samples, dye, and the single standard solution. The procedure for obtaining the gradients of the dye and standard solution is performed once at the beginning of analysis. The same procedure is applied thereafter for each sample. For illustration, the proposed automatic methodology was applied to determine total protein and albumin in blood serum by using the Biuret and Bromocresol Green (BCG) methods. The measurements were made by using a laboratory-made photometer based on a red and green bicolour LED (Light-Emitting Diode) and a phototransistor, coupled to a "Z" form flow cell. The sample throughput was about 50 h(-1) for albumin and 60 h(-1) for total protein, consuming about 7 microL of sample, 2.6 mL of BCG and 1.2 mL of biuret reagents for each determination. Applying the paired t-test for results from the proposed analyser and the reference method, no statistic differences at 95% confidence level were found. The absolute standard deviation was usually smaller than 0.2 g dL(-1). The proposed method is valuable for the determination of total protein and albumin; and can also be used in other determinations where kinetic effects may or may not exist.
Sigma-Aldrich
Bromocresol Green Sultone Form, for microscopy (Bot., Hist., Vit.), indicator (pH 3.8-5.4)
Sigma-Aldrich
Bromocresol Green, ACS reagent, Dye content 95 %