MilliporeSigma
  • Solvent-free synthesis of pyrimidine nucleoside-aminophosphonate hybrids and their biological activity evaluation.
Solvent-free synthesis of pyrimidine nucleoside-aminophosphonate hybrids and their biological activity evaluation.
Nucleosides, nucleotides & nucleic acids (2010-07-28)
Xinying Zhang, Yingying Qu, Xuesen Fan, C Bores, Dong Feng, Graciela Andrei, Robert Snoeck, Erik De Clercq, Philippe M Loiseau
ABSTRACT
A novel and highly efficient one-pot three-component synthesis of alpha-aminophosphonates under neat condition was developed. By employing this method, hybrid compounds of aminophosphonate with pyrimidine nucleosides were synthesized in good to excellent yields starting from 5-formyl-2'-deoxyuridine, aniline and dimethyl phosphite. The antiviral and antileishmanial activities of these novel hybrid compounds were also studied but none were found to be active.
Sigma-Aldrich
Dimethyl phosphite, 98%