MilliporeSigma
  • Synthesis and anti-HIV evaluation of new acyclic phosphonate nucleotide analogues and their bis(SATE) derivatives.
Synthesis and anti-HIV evaluation of new acyclic phosphonate nucleotide analogues and their bis(SATE) derivatives.
Nucleosides, nucleotides & nucleic acids (2010-07-28)
Hua Li, Joon Hee Hong
ABSTRACT
This article describes a very simple route for synthesizing novel lipophilic phosphonate bis(t-bu-SATE) prodrugs of acyclic cyclopentenylated nucleosides such as adenine 17 and cytosine 18. The key intermediate 6 was constructed via a ring-closing metathesis of compound 5, which could be readily prepared from diethylmalonate 4. The chemical stability of the bis(SATE) derivatives was tested at neutral (pH = 7.2) and slightly acid (milli-Q water, pH = 5.5) pH. The synthesized compounds were evaluated as potential antiviral agents against HIV-1 virus.
Sigma-Aldrich
Diethyl malonate, ReagentPlus®, 99%
Sigma-Aldrich
Diethyl malonate, ≥98%, FG