MilliporeSigma
  • Development and validation of a liquid chromatography-tandem mass spectrometry method for the determination of goserelin in rabbit plasma.
Development and validation of a liquid chromatography-tandem mass spectrometry method for the determination of goserelin in rabbit plasma.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2010-07-27)
Min Kyung Kim, Tae Ho Lee, Joon Hyuk Suh, Han Young Eom, Jung Won Min, Hyesun Yeom, Unyong Kim, Hyung Joon Jung, Kyung Hoi Cha, Yong Seok Choi, Jeong-Rok Youm, Sang Beom Han
ABSTRACT
A rapid and sensitive liquid chromatography-electrospray ionization tandem mass spectrometry method (LC-ESI-MS/MS) was developed and validated for the determination of goserelin in rabbit plasma. Various parameters affecting plasma sample preparation, LC separation, and MS/MS detection were investigated, and optimized conditions were identified. Acidified plasma samples were applied to Oasis((R)) HLB solid-phase extraction (SPE) cartridges. Extracted samples were evaporated under a stream of nitrogen and then reconstituted with 100microL mobile phase A. The separation was achieved on a Capcell-Pak C18 (2.0mmx150mm, 5microm, AQ type) column with a gradient elution of solvent A (0.05% acetic acid in deionized water/acetonitrile=85/15; v/v) and solvent B (acetonitrile) at a flow rate of 250microL/min. The LC-MS/MS system was equipped with an electrospray ion source operating in positive ion mode. Multiple-reaction monitoring (MRM) of the precursor-product ion transitions consisted of m/z 635.7-->m/z 607.5 for goserelin and m/z 424.0-->m/z 292.1 for cephapirin (internal standard). The proposed method was validated by assessing specificity, linearity, limit of quantification (LOQ), intra- and inter-day precision and accuracy, recovery, and stability. Linear calibration curves were obtained in the concentration range of 0.1-20ng/mL (the correlation coefficients were above 0.99). The LOQ of the method was 0.1ng/mL. Results obtained from the validation study of goserelin showed good accuracy and precision at concentrations of 0.1, 1, 5, 10, and 20ng/mL. The validated method was successfully applied to a pharmacokinetic study of goserelin after a single subcutaneous injection of 3.6mg of goserelin in healthy white rabbits.
Product Description
Supelco
Cefapirin sodium, analytical standard