MilliporeSigma
  • Rapid photo-crosslinking of fumaric acid monoethyl ester-functionalized poly(trimethylene carbonate) oligomers for drug delivery applications.
Rapid photo-crosslinking of fumaric acid monoethyl ester-functionalized poly(trimethylene carbonate) oligomers for drug delivery applications.
Journal of controlled release : official journal of the Controlled Release Society (2010-07-21)
Janine Jansen, Mark J Boerakker, Jean Heuts, Jan Feijen, Dirk W Grijpma
ABSTRACT
Photo-crosslinkable, fumaric acid monoethyl ester-functionalized poly(trimethylene carbonate) oligomers were synthesized and copolymerized with N-vinyl pyrrolidone (NVP) and vinyl acetate (VAc) to form biodegradable polymer networks. The copolymerization reactions were much faster than homopolymerization of the fumarate end-groups of the macromers. The hydrophilicity of the networks could by varied by mixing NVP and VAc at different ratios. The prepared network extracts were compatible with NIH 3T3 fibroblasts. Release of vitamin B12, used as a model drug, could be tuned by varying network hydrophilicity and macromer molecular weight. A more hydrophilic and less densely crosslinked network resulted in faster release.
Product Description
Supelco
Vinyl acetate solution, certified reference material, 5000 μg/mL in acetonitrile
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 g
Estimated to ship on March 21, 2025
This SKU is also available as ACS grade in our Exclusive Redi-Dri™ free-flowing product line.
US$260.00
1 kg
Estimated to ship on March 21, 2025
This SKU is also available as ACS grade in our Exclusive Redi-Dri™ free-flowing product line.
US$371.00
2.5 kg
Estimated to ship on March 21, 2025
This SKU is also available as ACS grade in our Exclusive Redi-Dri™ free-flowing product line.
US$655.00
12 kg
Estimated to ship on March 21, 2025
This SKU is also available as ACS grade in our Exclusive Redi-Dri™ free-flowing product line.
US$1,490.00
Sigma-Aldrich
mono-Ethyl fumarate, 95%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 g
Estimated to ship on March 21, 2025
This SKU is also available as ACS grade in our Exclusive Redi-Dri™ free-flowing product line.
US$260.00
1 kg
Estimated to ship on March 21, 2025
This SKU is also available as ACS grade in our Exclusive Redi-Dri™ free-flowing product line.
US$371.00
2.5 kg
Estimated to ship on March 21, 2025
This SKU is also available as ACS grade in our Exclusive Redi-Dri™ free-flowing product line.
US$655.00
12 kg
Estimated to ship on March 21, 2025
This SKU is also available as ACS grade in our Exclusive Redi-Dri™ free-flowing product line.
US$1,490.00
Sigma-Aldrich
1-Vinyl-2-pyrrolidinone, contains sodium hydroxide as inhibitor, ≥99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 g
Estimated to ship on March 21, 2025
This SKU is also available as ACS grade in our Exclusive Redi-Dri™ free-flowing product line.
US$260.00
1 kg
Estimated to ship on March 21, 2025
This SKU is also available as ACS grade in our Exclusive Redi-Dri™ free-flowing product line.
US$371.00
2.5 kg
Estimated to ship on March 21, 2025
This SKU is also available as ACS grade in our Exclusive Redi-Dri™ free-flowing product line.
US$655.00
12 kg
Estimated to ship on March 21, 2025
This SKU is also available as ACS grade in our Exclusive Redi-Dri™ free-flowing product line.
US$1,490.00
Sigma-Aldrich
Vinyl acetate, contains 3-20 ppm hydroquinone as inhibitor, ≥99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 g
Estimated to ship on March 21, 2025
This SKU is also available as ACS grade in our Exclusive Redi-Dri™ free-flowing product line.
US$260.00
1 kg
Estimated to ship on March 21, 2025
This SKU is also available as ACS grade in our Exclusive Redi-Dri™ free-flowing product line.
US$371.00
2.5 kg
Estimated to ship on March 21, 2025
This SKU is also available as ACS grade in our Exclusive Redi-Dri™ free-flowing product line.
US$655.00
12 kg
Estimated to ship on March 21, 2025
This SKU is also available as ACS grade in our Exclusive Redi-Dri™ free-flowing product line.
US$1,490.00
Vinyl acetate, European Pharmacopoeia (EP) Reference Standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 g
Estimated to ship on March 21, 2025
This SKU is also available as ACS grade in our Exclusive Redi-Dri™ free-flowing product line.
US$260.00
1 kg
Estimated to ship on March 21, 2025
This SKU is also available as ACS grade in our Exclusive Redi-Dri™ free-flowing product line.
US$371.00
2.5 kg
Estimated to ship on March 21, 2025
This SKU is also available as ACS grade in our Exclusive Redi-Dri™ free-flowing product line.
US$655.00
12 kg
Estimated to ship on March 21, 2025
This SKU is also available as ACS grade in our Exclusive Redi-Dri™ free-flowing product line.
US$1,490.00
Supelco
Vinyl acetate, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 g
Estimated to ship on March 21, 2025
This SKU is also available as ACS grade in our Exclusive Redi-Dri™ free-flowing product line.
US$260.00
1 kg
Estimated to ship on March 21, 2025
This SKU is also available as ACS grade in our Exclusive Redi-Dri™ free-flowing product line.
US$371.00
2.5 kg
Estimated to ship on March 21, 2025
This SKU is also available as ACS grade in our Exclusive Redi-Dri™ free-flowing product line.
US$655.00
12 kg
Estimated to ship on March 21, 2025
This SKU is also available as ACS grade in our Exclusive Redi-Dri™ free-flowing product line.
US$1,490.00