MilliporeSigma
  • Therapeutic effects of vitamin D analogs on cardiac hypertrophy in spontaneously hypertensive rats.
Therapeutic effects of vitamin D analogs on cardiac hypertrophy in spontaneously hypertensive rats.
The American journal of pathology (2010-07-10)
Juan Kong, Gene H Kim, Minjie Wei, Tao Sun, George Li, Shu Q Liu, Xinmin Li, Ishir Bhan, Qun Zhao, Ravi Thadhani, Yan Chun Li
ABSTRACT
Vitamin D inhibits renin expression and blocks the compensatory induction of renin associated with the use of renin-angiotensin system inhibitors. Here we test the therapeutic effects of two commonly used vitamin D analogs and their combination with losartan on the development of left ventricular hypertrophy. One-month-old male spontaneously hypertensive rats were treated with vehicle, losartan, paricalcitol, doxercalciferol, a combination of losartan and paricalcitol, or a combination of losartan and doxercalciferol for 2 months. Blood pressure was markedly reduced by losartan, but not by paricalcitol or doxercalciferol alone. Echocardiograpy demonstrated a 65 to 80% reduction in left ventricular wall thickness with losartan, paricalcitol, or doxercalciferol monotherapy and almost complete prevention of left ventricular hypertrophy with the combination therapies. Attenuation of cardiac and cardiomyocyte hypertrophy, and suppression of atrial and brain natriuretic peptides, were most marked in the combination therapy groups. These changes were well correlated with left ventricular gene and microRNA expression profiles in the different treatment groups. Renal and cardiac renin expression was markedly increased in losartan-treated animals, but nearly normalized with combination therapy. The same vitamin D analogs suppressed plasma renin activity in patients receiving chronic hemodialysis. These data demonstrate that vitamin D analogs have potent antihypertrophic activity in part via suppression of renin in the kidney and heart, and combination of these analogs with losartan achieves much better therapeutic effects because of the blockade of the compensatory renin increase.
Sigma-Aldrich
Doxercalciferol, ≥98% (HPLC), solubility: >10 mg/mL in DMSO