MilliporeSigma
  • Antibody functionalized interdigitated micro-electrode (IDmicroE) based impedimetric cortisol biosensor.
Antibody functionalized interdigitated micro-electrode (IDmicroE) based impedimetric cortisol biosensor.
The Analyst (2010-07-01)
Sunil K Arya, Ganna Chornokur, Manju Venugopal, Shekhar Bhansali
ABSTRACT
This paper reports on an ultrasensitive, disposable, impedimetric biosensor for cortisol detection. C-Mab (a Cortisol specific monoclonal antibody) was covalently immobilized via amide bond on the surface of the interdigitated micro-electrodes (IDmicroEs) functionalized with dithiobis(succinimidyl propionate) (DTSP) self-assembled monolayer (SAM). After C-Mab binding, unreacted active groups of DTSP were blocked using ethanolamine (EA) and glycine (Gly) mixture. The disposable sensors were exposed to solutions with different cortisol concentrations and a label-free electrochemical impedance (EIS) technique was used to determine the cortisol concentration. EIS results confirm that the EA-Gly/C-Mab/DTSP/IDmicroE based biosensor exhibited the sensitivity of 2.855 kohms M(-1) and could accurately detect cortisol in the range of 1 pM to 10 nM in saliva. This work establishes the feasibility of using an impedance based biosensor as a disposable cortisol detector, capable of working with complex bodily fluids (e.g., saliva). The architecture enables the use of cortisol sensors at point-of-care.
Sigma-Aldrich
3,3′-Dithiodipropionic acid di(N-hydroxysuccinimide ester), powder