MilliporeSigma
  • Nitrogen-doped TiO2 modified with NH4F for efficient photocatalytic degradation of formaldehyde under blue light-emitting diodes.
Nitrogen-doped TiO2 modified with NH4F for efficient photocatalytic degradation of formaldehyde under blue light-emitting diodes.
Journal of hazardous materials (2010-06-29)
Yuexiang Li, Yuan Jiang, Shaoqin Peng, Fengyi Jiang
ABSTRACT
A nitrogen-doped TiO(2) (N-TiO(2)) photocatalyst was prepared by calcination of the hydrolysis precipitate of Ti(SO(4))(2) with aqueous ammonia. The prepared N-TiO(2) was treated with NH(4)F (F-N-TiO(2)) by an impregnation-calcination method. The photocatalyst (F-N-TiO(2)) was characterized by X-ray diffraction (XRD), Fourier Transform Infrared (FT-IR), UV-vis diffusive reflectance spectroscopy (DRS), BET and X-ray photoelectron spectroscopy (XPS). With blue light-emitting diode (LED) as the light source, its photocatalytic activity for the degradation of formaldehyde was investigated. NH(4)F treatment enhances markedly photocatalytic activity of N-TiO(2). The treatment increases the visible absorption of N-TiO(2), decreases its specific surface area and influences the concentration of oxygen vacancies in N-TiO(2). Photocatalytic activity of F-N-TiO(2) depends on the visible absorption, the specific surface area, and the concentration of oxygen vacancies. The preparation conditions, such as the calcination temperature and the initial molar ratio of NH(4)F to N-TiO(2), have a significant influence on the photocatalytic activity. The doping mechanism of NH(4)F was investigated.
Sigma-Aldrich
Ammonium hydrogen difluoride, 99.999% trace metals basis
Sigma-Aldrich
Ammonium hydrogen difluoride, reagent grade, 95%
Sigma-Aldrich
Ammonium fluoride, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium fluoride, ACS reagent, ≥98.0%