MilliporeSigma
  • Evaluation of three chitin metal silicate co-precipitates as a potential multifunctional single excipient in tablet formulations.
Evaluation of three chitin metal silicate co-precipitates as a potential multifunctional single excipient in tablet formulations.
Marine drugs (2010-06-19)
Rana Al-Shaikh Hamid, Faisal Al-Akayleh, Mohammad Shubair, Iyad Rashid, Mayyas Al Remawi, Adnan Badwan
ABSTRACT
The performance of the novel chitin metal silicate (CMS) co-precipitates as a single multifunctional excipient in tablet formulation using direct compression and wet granulation methods is evaluated. The neutral, acidic, and basic drugs Spironolactone (SPL), ibuprofen (IBU) and metronidazole (MET), respectively, were used as model drugs. Commercial Aldactone, Fleximex and Dumazole tablets containing SPL, IBU and MET, respectively, and tablets made using Avicel 200, were used in the study for comparison purposes. Tablets of acceptable crushing strength (>40 N) were obtained using CMS. The friability values for all tablets were well below the maximum 1% USP tolerance limit. CMS produced superdisintegrating tablets (disintegration time < 1 min) with the three model drugs. Regarding the dissolution rate, the sequence was as follow: CMS > Fleximex > Avicel 200, CMS > Avicel 200 > Dumazole and Aldactone > Avicel 200 > CMS for IBU, MET and SPL, respectively. Compressional properties of formulations were analyzed using density measurements and the compression Kawakita equation as assessment parameters. On the basis of DSC results, CMS co precipitates were found to be compatible with the tested drugs. Conclusively, the CMS co-precipitates have the potential to be used as filler, binder, and superdisintegrant, all-in-one, in the design of tablets by the direct compression as well as wet granulation methods.
Product Description
Supelco
Florisil® Adsorbent for Chromatography, 30-60 mesh
Supelco
Florisil® Adsorbent for Chromatography, 60-100 mesh
Supelco
Florisil®, 60-100 mesh, coarse powder
Supelco
Florisil®, 100-200 mesh, fine powder
Supelco
Florisil®, for the determination of hydrocarbon acc. to ISO 9377-2
Supelco
Florisil®, <200 mesh, fine powder
Supelco
Florisil® PR
Sigma-Aldrich
Florisil®, 100-200 mesh