MilliporeSigma
  • Therapeutic evaluation of free and liposome-loaded furazolidone in experimental visceral leishmaniasis.
Therapeutic evaluation of free and liposome-loaded furazolidone in experimental visceral leishmaniasis.
International journal of antimicrobial agents (2010-06-18)
André Gustavo Tempone, Renato Arruda Mortara, Heitor Franco de Andrade, Juliana Quero Reimão
ABSTRACT
Drug delivery systems are promising pharmaceutical formulations used to improve the therapeutic index of drugs. In this study, we developed a liposomal formulation of furazolidone that targets Leishmania (Leishmania) chagasi amastigotes in a hamster model. Using laser scanning confocal microscopy, it was demonstrated that the liposomal drug co-localised with L. (L.) chagasi amastigotes within macrophages. Liposomal furazolidone administered intraperitoneally at 0.5mg/kg for 12 consecutive days reduced spleen (74%) and liver (32%) parasite burden at a 100-fold lower dose than the free drug. Free furazolidone (50mg/kg) also effectively reduced spleen (82.5%) and liver (85%) parasites; its in vitro activity against promastigotes and intracellular amastigotes demonstrated a high degree of parasite selectivity. Thus, furazolidone, both in the free and liposome-loaded formulation, is an effective inhibitor of L. (L.) chagasi, representing a possible cost-effective drug candidate for the treatment of visceral leishmaniasis.
Sigma-Aldrich
Furazolidone