MilliporeSigma
  • Synthesis and in vivo evaluation of [11C]MPTQ: a potential PET tracer for alpha2A-adrenergic receptors.
Synthesis and in vivo evaluation of [11C]MPTQ: a potential PET tracer for alpha2A-adrenergic receptors.
Bioorganic & medicinal chemistry letters (2010-06-10)
Jaya Prabhakaran, Vattoly J Majo, Matthew S Milak, Pratap Mali, Lyudmila Savenkova, J John Mann, Ramin V Parsey, J S Dileep Kumar
ABSTRACT
Radiosynthesis and in vivo evaluation of [N-methyl-(11)C] 5-methyl-3-[4-(3-phenylallyl)-piperazin-1-ylmethyl]-3,3a,4,5-tetrahydroisoxazolo[4,3-c]quinoline (1), a potential PET tracer for alpha2-adrenergic receptors is described. Syntheses of nonradioactive standard 1 and corresponding desmethyl precursor 2 were achieved from 2-aminobenzaldehyde in 40% and 65% yields, respectively. Methylation using [(11)C]CH(3)I in presence of aqueous potassium hydroxide in DMSO afforded [(11)C]1 in 25% yield (EOS) with >99% chemical and radiochemical purities with a specific activity ranged from 3-4 Ci/micromol (n=6). The total synthesis time was 30 min from EOB. PET studies in anesthetized baboon show that [(11)C]1 penetrates BBB and accumulates in alpha2A-AR enriched brain areas.
Sigma-Aldrich
2-Aminobenzaldehyde, ≥98%