MilliporeSigma
  • Development of an enzyme-linked immunosorbent assay specific to Sudan red I.
Development of an enzyme-linked immunosorbent assay specific to Sudan red I.
Analytical biochemistry (2010-06-05)
Ting Xu, Ke Yi Wei, Jia Wang, Sergei A Eremin, Shang Zhong Liu, Qing X Li, Ji Li
ABSTRACT
To obtain antibodies to develop an enzyme-linked immunosorbent assay (ELISA) for the analysis of Sudan red I, haptens were designed and synthesized via four different strategies: (i) attachment of a spacer at the para position of the benzene ring, (ii) attachment of a spacer at the naphthol part, (iii) attachment of a spacer at the hydroxyl group of the Sudan red I molecule, and (iv) use of a fragment of the target molecule. A total of 10 haptens were used to generate immunogens, coating antigens, and polyclonal antibodies. One of the heterologous ELISAs developed exhibited an IC(50) of 1.6 ng/ml, a limit of detection (LOD) of 0.03 ng/ml, and a dynamic range between 0.1 and 14 ng/ml. The assay had 13% cross-reactivity with Para red and negligible cross-reactivity with other structure-related compounds. This ELISA was much more specific than those published previously. This assay was used to determine Sudan red I residues in tomato sauce and chili powder samples after simple pretreatment. The results were validated by comparison with high-performance liquid chromatography (HPLC). The average recoveries of Sudan red I by ELISA and HPLC were in ranges of 70-97% and 82-114%, respectively, indicating suitability of the developed ELISA for screening of Sudan red I in foods.
Product Description
Supelco
Sudan II, analytical standard
Sigma-Aldrich
Sudan II, Dye content 90 %