MilliporeSigma
  • First representative of optically active P-L-menthyl-substituted (aminomethyl)phosphine and its borane and metal complexes.
First representative of optically active P-L-menthyl-substituted (aminomethyl)phosphine and its borane and metal complexes.
Inorganic chemistry (2010-05-28)
Svetlana N Ignatieva, Anna S Balueva, Andrey A Karasik, Shamil K Latypov, Anna G Nikonova, Olga E Naumova, Peter Lönnecke, Evamarie Hey-Hawkins, Oleg G Sinyashin
ABSTRACT
The first representative of 1,5-diaza-3,7-diphosphacyclooctanes (1) with chiral L-menthyl substituents on the phosphorus atoms was obtained by condensation of L-menthylphosphine with formaldehyde and p-toluidine. This optically active cyclic bisphosphine readily forms a stable P,P-complex with borane (2) and P,P-chelate complexes with platinum(II) (3) and palladium(II) dichloride (4). The structure of the bisphosphine 1 in solution was elucidated by employing a variety of 1D/2D NMR correlation experiments, and the molecular structure of complex 3 was studied by X-ray crystallography.
Sigma-Aldrich
p-Toluidine hydrochloride, purum, ≥99.0% (AT)
Supelco
p-Toluidine, for spectrophotometric det. of Au, Tl(III), W, ≥99.0%
Sigma-Aldrich
p-Toluidine, 99%
Sigma-Aldrich
p-Toluidine, 99.6%