MilliporeSigma
  • Unconventional U-shaped conformation of a bolaamphiphile embedded in a synthetic host.
Unconventional U-shaped conformation of a bolaamphiphile embedded in a synthetic host.
Chemical communications (Cambridge, England) (2010-05-19)
Kangkyun Baek, Youngkook Kim, Hyunuk Kim, Minyoung Yoon, Ilha Hwang, Young Ho Ko, Kimoon Kim
ABSTRACT
The X-ray crystal structure of 1,12-dodecane diammonium (C(12)DA(2+)) encapsulated in cucurbit[8]uril reveals an unconventional U-shaped conformation of C(12)DA(2+), which is attributed to the favorable host-guest interactions to overcome the charge-charge repulsion of the two ammonium groups in close proximity and high internal strain.
Sigma-Aldrich
1,12-Diaminododecane, 98%