MilliporeSigma
  • General route toward side-chain-functionalized alpha-helical polypeptides.
General route toward side-chain-functionalized alpha-helical polypeptides.
Biomacromolecules (2010-05-15)
Haoyu Tang, Donghui Zhang
ABSTRACT
A series of poly(gamma-chloropropyl-L-glutamate) (PCPLG) with controlled polymer molecular weight (MW = 5-28 kg x mol(-1)) and molecular weight distribution (PDI = 1.16-1.26) have been prepared from hexamethyldisilazane (HMDS)-mediated ring-opening polymerization (ROP) of gamma-chloropropyl-L-glutamic acid based N-carboxylanhydride (CP-NCA). CD, FTIR, and WAXS analysis reveal that the polymers adopt alpha-helical conformations both in solution and the solid state. Their helical surfaces can be readily decorated with functional moieties to confer desired properties, as demonstrated by the quantitative derivatization of the PCPLG side chains with azido functional groups that are amendable to "click" chemistry. Subsequent side-chain conjugation with mannose moieties via copper-mediated [2 + 3] alkyne-azide 1,3-dipolar cycloaddition affords water-soluble mannose-polypeptide conjugates with quantitative grafting efficiency occurring under mild conditions. CD analysis reveals that the mannose-polypeptide conjugates also retain alpha-helical conformations in aqueous solution.
Sigma-Aldrich
Potassium bis(trimethylsilyl)amide solution, 1 M in THF
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide solution, 1 M in toluene
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide solution, 1 M in tert-butyl methyl ether
Sigma-Aldrich
Potassium bis(trimethylsilyl)amide solution, 0.5 M in toluene
Sigma-Aldrich
Hexamethyldisilazane, reagent grade, ≥99%
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide solution, 1.0 M in THF
Supelco
Hexamethyldisilazane, for GC derivatization, LiChropur, ≥99.0% (GC)
Sigma-Aldrich
Hexamethyldisilazane, Wacker Chemie AG, ≥97.0% (GC)
Sigma-Aldrich
Hexamethyldisilazane, ReagentPlus®, 99.9%
Sigma-Aldrich
Sodium bis(trimethylsilyl)amide solution, 1.0 M in THF
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide solution, 1.0 M in hexanes
Sigma-Aldrich
Sodium bis(trimethylsilyl)amide solution, 0.6 M in toluene
Sigma-Aldrich
Sodium bis(trimethylsilyl)amide solution, 40% in THF
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide solution, 1.5 M in THF
Sigma-Aldrich
Potassium bis(trimethylsilyl)amide solution, 1.0 M in 2-methyltetrahydrofuran
Sigma-Aldrich
Sodium bis(trimethylsilyl)amide, 95%
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide solution, 0.5 M in 2-methyltetrahydrofuran
Sigma-Aldrich
Potassium bis(trimethylsilyl)amide, 95%
Sigma-Aldrich
Tris[N,N-bis(trimethylsilyl)amide]yttrium
Sigma-Aldrich
Tris[N,N-bis(trimethylsilyl)amide]samarium(III), 98%
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide, 97%