MilliporeSigma
  • Development and validation of a dissolution test method for vitamin A in dietary supplement tablets.
Development and validation of a dissolution test method for vitamin A in dietary supplement tablets.
Journal of pharmaceutical and biomedical analysis (2010-04-30)
Natalia Davydova, Erika Stippler, Ping Jin, Gabriel Giancaspro
ABSTRACT
A dissolution test method and an analytical procedure by HPLC were developed and validated for evaluation of the dissolution behavior of dietary supplements tablets containing vitamin A in the forms of retinyl acetate or retinyl palmitate. Seven different commercially available products containing retinyl acetate or retinyl palmitate were selected for this study. A dissolution medium containing 1% (w/v) Octoxynol 9 (Triton X-100) and 1% (w/v) (+)-sodium alpha-ascorbate in 0.05 M phosphate buffer, pH 6.8, was found suitable to ensure sink conditions and chemical stability for both retinyl acetate and retinyl palmitate. Two rotation speeds, 50 and 75 rpm, were evaluated with USP Apparatus 2 and 900 ml dissolution medium. Dissolution profiles were generated over 120 min. Dissolution samples were analyzed with a reversed-phase HPLC method with UV detection at 325 nm. Each product was also assayed for vitamin A content according to USP 32-NF 27. The results from 45 min to the last time point of the dissolution tests performed at 75 rpm were consistent with the Assay results. The dissolution test described here could be proposed as a pharmacopeial standard to assess the performance of tablet formulations containing vitamin A as retinyl esters.
Product Description
Supelco
Retinyl acetate, analytical standard
Sigma-Aldrich
Retinyl acetate, solid or viscous liquid, BioReagent, synthetic, suitable for cell culture
Sigma-Aldrich
Retinyl acetate, synthetic, crystalline solid or supercooled liquid
Sigma-Aldrich
Retinyl acetate, synthetic, matrix dispersion, 475,000-650,000 USP units/g
Retinol acetate, European Pharmacopoeia (EP) Reference Standard
Supelco
Retinyl Acetate (Vitamin A Acetate), Pharmaceutical Secondary Standard; Certified Reference Material