MilliporeSigma
  • Syringin from stem bark of Fraxinus rhynchophylla protects Abeta(25-35)-induced toxicity in neuronal cells.
Syringin from stem bark of Fraxinus rhynchophylla protects Abeta(25-35)-induced toxicity in neuronal cells.
Archives of pharmacal research (2010-04-28)
Eun-Ju Yang, Sang-In Kim, Hyun-Yeong Ku, Dong-Seok Lee, Jong-Won Lee, Yae-Sil Kim, Yeon-Hee Seong, Kyung-Sik Song
ABSTRACT
The medicinal herb Jinpi, derived from the dried stem barks of Fraxinus rhynchophylla belonging to Oleaceae is widely used as a variety of Korean folk remedies for anti-inflammatory, febricide, antidiarrhea, and antileukorrhea diseases. In the course of screening antidementia agents from natural products, F. rhynchophylla showed significant inhibitory activity toward Abeta(25-35)-induced neuronal cell death. An active principle was isolated and identified as syringin. When the neuroblastoma cells were exposed to 50 microM Abeta(25-35), 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) reduction rate (survival rate) decreased to 60.21 +/- 2.16% over control while syringin treated ones recovered cell viability up to 79.12 +/- 1.39% at 20 microM. In addition, 20 microM syringin almost completely removed Abeta(25-35)-induced reactive oxygen species. The neuroprotective effect of syringin seemed to be originated from the reduction of apoptosis since decrease in caspase-3 activity and expression, reduction in cleaved PARP, and DNA fragmentation were observed. These results suggest that F. rhynchophylla and syringin are expected to be useful for preventing Abeta(25-35)-induced neuronal cell damage.
Product Description
Supelco
Eleutheroside B, analytical standard