MilliporeSigma
  • Assessment of bile fluorescence patterns in a tropical fish, Nile tilapia (Oreochromis niloticus) exposed to naphthalene, phenanthrene, pyrene and chrysene using fixed wavelength fluorescence and synchronous fluorescence spectrometry.
Assessment of bile fluorescence patterns in a tropical fish, Nile tilapia (Oreochromis niloticus) exposed to naphthalene, phenanthrene, pyrene and chrysene using fixed wavelength fluorescence and synchronous fluorescence spectrometry.
Bulletin of environmental contamination and toxicology (2010-04-23)
A Pathiratne, C K Hemachandra, K A S Pathiratne
ABSTRACT
Bile fluorescence patterns in Nile tilapia, a potential fish for biomonitoring tropical water pollution were assessed following exposure to selected polycyclic aromatic hydrocarbons (PAHs): naphthalene, phenanthrene, pyrene and chrysene. Non-normalized fixed wavelength fluorescence signals in the fish exposed to these PAHs reflected dose and/or time response relationships of their metabolism. Normalizing signals to biliverdin introduced deviations to these response patterns. The optimal wavelength pairs (excitation/emission) for synchronous fluorescence scanning measurements of bile metabolites of naphthalene, phenanthrene, pyrene and chrysene were identified as 284/326, 252/357, 340/382 and 273/382 respectively. This study supports the use of bile fluorescence in Nile tilapia by fixed wavelength fluorescence and synchronous fluorescence spectrometry with non-normalized data as a simple method for screening bioavailability of these PAHs.
Product Description
Supelco
Chrysene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Chrysene, analytical standard
Chrysene, BCR®, certified reference material