MilliporeSigma
A chiral solvent effect in asymmetric organocatalysis.
Organic letters (2010-04-20)
Michael North, Pedro Villuendas
ABSTRACT
Proline-catalyzed aldol reactions between enolizable ketones and aromatic aldehydes can be carried out in propylene carbonate. When enantiomerically pure propylene carbonate is used, the combination of (R)-proline and (R)-propylene carbonate constitutes a matched pair, while (S)-proline and (R)-propylene carbonate constitutes a mismatched pair.
Sigma-Aldrich
Propylene carbonate, anhydrous, 99.7%
Sigma-Aldrich
Propylene carbonate, ReagentPlus®, 99%
Sigma-Aldrich
Propylene carbonate, suitable for HPLC, 99.7%