MilliporeSigma
  • Histopathologic validation of 3'-deoxy-3'-18F-fluorothymidine PET in squamous cell carcinoma of the oral cavity.
Histopathologic validation of 3'-deoxy-3'-18F-fluorothymidine PET in squamous cell carcinoma of the oral cavity.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine (2010-04-17)
Esther G C Troost, Johan Bussink, Piet J Slootweg, Wenny J M Peeters, Matthias A W Merkx, Albert J van der Kogel, Wim J G Oyen, Johannes H A M Kaanders
ABSTRACT
Accelerated tumor cell repopulation is an important mechanism adversely affecting therapeutic outcome in head and neck cancer. The noninvasive assessment of the proliferative state of a tumor by PET may provide a selection tool for customized treatment. 3'-deoxy-3'-(18)F-fluorothymidine ((18)F-FLT) is a PET tracer that is phosphorylated by thymidine kinase 1 (TK-1) and, as such, reflects cellular proliferation. Before the use of (18)F-FLT PET for tumor characterization is accepted and introduced into clinical studies, validation against tumor histology is mandatory. The aim of this study was to validate (18)F-FLT PET in squamous cell carcinomas of the oral cavity using immunohistochemical staining for the proliferation marker iododeoxyuridine and for TK-1. Seventeen patients with primary squamous cell carcinomas of the oral cavity underwent an (18)F-FLT PET/CT scan before surgery, and iododeoxyuridine was administered 20 min before tumor resection. (18)F-FLT PET/CT scans were segmented, and PET/CT volumes and PET signal intensities were calculated (mean standardized uptake value [SUV(mean)] and maximum standardized uptake value [SUV(max)]). Multiple paraffin-embedded tumor sections were immunohistochemically stained for iododeoxyuridine and TK-1. For iododeoxyuridine, labeling indices and optical densities were calculated and correlated with SUV(mean) and SUV(max). TK-1 staining was visually and semiquantitatively assessed. All primary tumors were identified with (18)F-FLT PET but with a large range in tracer uptake (mean SUV(max), 5.9; range, 2.2-15.2). Also, there was a large variability in iododeoxyuridine labeling indices (mean, 0.09; range, 0.01-0.29) and optical densities (mean, 28.2; range, 12.6-37.8). The iododeoxyuridine optical densities correlated significantly with SUV(mean) and SUV(max), but the labeling indices did not. In most tumors, TK-1 staining of varying intensity was present but correlated with neither iododeoxyuridine binding nor (18)F-FLT uptake. The current study demonstrated only a weak correlation between (18)F-FLT uptake and iododeoxyuridine staining intensity in oral cavity tumors. This weak correlation may be explained by differences in biomarker characteristics, resolution, and quantification methods.
Sigma-Aldrich
5-Iodo-2′-deoxyuridine, ≥99% (HPLC)