MilliporeSigma
  • A practical and azide-free synthetic approach to oseltamivir from diethyl D-tartrate.
A practical and azide-free synthetic approach to oseltamivir from diethyl D-tartrate.
The Journal of organic chemistry (2010-04-17)
Jiang Weng, Yong-Bo Li, Rui-Bin Wang, Feng-Quan Li, Can Liu, Albert S C Chan, Gui Lu
ABSTRACT
A short and practical synthesis of oseltamivir was accomplished in 11 steps from inexpensive and abundant diethyl D-tartrate starting material. This azide-free route featured an asymmetric aza-Henry reaction and a domino nitro-Michael/Horner-Wadsworth-Emmons (HWE) reaction as the key steps to construct the relevant cyclohexene ring of the product, which provided an economical and practical alternative for the synthesis of oseltamivir.
Sigma-Aldrich
(+)-Diethyl L-tartrate, ≥99%
Sigma-Aldrich
Diethyl L-tartrate, ≥99%, FG
Sigma-Aldrich
(−)-Diethyl D-tartrate, ≥99%