MilliporeSigma
  • Evaluation of 111In-labeled cyclic RGD peptides: tetrameric not tetravalent.
Evaluation of 111In-labeled cyclic RGD peptides: tetrameric not tetravalent.
Bioconjugate chemistry (2010-04-15)
Sudipta Chakraborty, Jiyun Shi, Young-Seung Kim, Yang Zhou, Bing Jia, Fan Wang, Shuang Liu
ABSTRACT
This report presents the synthesis and evaluation of (111)In(DOTA-6G-RGD(4)) (DOTA = 1,4,7,10-tetraazacyclododecane-1,4,7,10-tetracetic acid; 6G-RGD(4) = E{G(3)-E[G(3)-c(RGDfK)](2)}(2) and G(3) = Gly-Gly-Gly), (111)In(DOTA-RGD(4)) (RGD(4) = E{E[c(RGDfK)](2)}(2)) and (111)In(DOTA-3G-RGD(2)) (3G-RGD(2) = G(3)-E[G(3)-c(RGDfK)](2)) as new radiotracers for imaging integrin alpha(v)beta(3)-positive tumors. The IC(50) values of DOTA-6G-RGD(4), DOTA-RGD(4), and DOTA-3G-RGD(2) were determined to be 0.4 +/- 0.1, 1.4 +/- 0.1 and 1.1 +/- 0.1 nM against (125)I-c(RGDyK) bound to integrin alpha(v)beta(3)-positive U87MG human glioma cells. (111)In(DOTA-6G-RGD(4)), (111)In(DOTA-RGD(4)), and (111)In(DOTA-3G-RGD(2)) were prepared by reacting (111)InCl(3) with the respective DOTA conjugate in NH(4)OAc buffer (100 mM, pH = 5.5). Radiolabeling could be completed by heating the reaction mixture at 100 degrees C for 15-20 min. The specific activity was approximately 1850 MBq/micromol for (111)In(DOTA-3G-RGD(2)) and approximately 1480 MBq/micromol for (111)In(DOTA-6G-RGD(4)). The athymic nude mice bearing U87MG human glioma xenografts were used to evaluate tumor uptake and excretion kinetics of (111)In(DOTA-6G-RGD(4)), (111)In(DOTA-RGD(4)), and (111)In(DOTA-3G-RGD(2)). The results from both the integrin alpha(v)beta(3) binding assay and biodistribution studies suggest that the tetrameric cyclic RGD peptides, such as RGD(4) and 6G-RGD(4), are most likely bivalent in binding to the integrin alpha(v)beta(3). Both (111)In(DOTA-6G-RGD(4)) and (111)In(DOTA-RGD(4)) had significantly higher tumor uptake than (111)In(DOTA-3G-RGD(2)) at 24-72 h postinjection due to the extra RGD motifs in RGD(4) and 6G-RGD(4). (111)In(DOTA-3G-RGD(2)) had very little metabolism, while (111)In(DOTA-6G-RGD(4)) had significant metabolism during its excretion via both renal and hepatobiliary routes over the 2 h period, probably due to its much larger size. The combination of high tumor uptake with long tumor retention suggests that their corresponding (90)Y and (177)Lu analogues M(DOTA-6G-RGD(4)) (M = (90)Y and (177)Lu) might be useful as therapeutic radiotracers for treatment of integrin alpha(v)beta(3)-positive solid tumors.
Sigma-Aldrich
Gly-Gly-Gly, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Gly-Gly-Gly