MilliporeSigma
  • Synthesis of thermo-sensitive copolymer with affinity butyl ligand and its application in lipase purification.
Synthesis of thermo-sensitive copolymer with affinity butyl ligand and its application in lipase purification.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2010-04-07)
Libo Zhou, Junfen Wan, Xuejun Cao
ABSTRACT
In this study, thermo-sensitive N-alkyl substituted polyacrylamide polymer P(NNB) was synthesized by using N-hydroxymethyl acrylamide(NHAM), N-isopropyl acrylamide (NIPA) and butyl acrylate (BA) as monomers, and its low critical solution temperature (LCST) was controlled to be 28 degrees C. The recovery of the thermo-sensitive polymer was over 98%. Butanol as a hydrophobic ligand was covalently attached onto polymer P(NNB) and butyl ligand density was 80 micromol g(-1) polymer. The affinity polymer was used for purification of lipase from crude material. Optimized condition was pH 7.0, 35 degrees C adsorption temperature, 120 min adsorption time and 0.5 mg ml(-1) initial concentration of lipase. The adsorption isotherm accords with a typical Langmuir isotherm. The maximum adsorption capacity (Q(m)) of the affinity polymer for lipase was 24.8 mg g(-1)polymer. The affinity copolymer could be recycled by temperature-inducing precipitation and there was only about 6% loss of adsorption capacity after five recyclings. Specific activity of lipase was improved from 14IU mg(-1) to 506 IU mg(-1) protein, and its recovery achieved 82%. The affinity polymer is suitable for the purification of target proteins from the crude material with large volume and dilute solution.
Sigma-Aldrich
N-(Hydroxymethyl)acrylamide solution, 48 wt. % in H2O
Supelco
Butyl acrylate, analytical standard
Sigma-Aldrich
Butyl acrylate, ≥99%, contains 10-60 ppm monomethyl ether hydroquinone as inhibitor