MilliporeSigma
  • Microbial transformation of ginsenosides Rb1, Rb3 and Rc by Fusarium sacchari.
Microbial transformation of ginsenosides Rb1, Rb3 and Rc by Fusarium sacchari.
Journal of applied microbiology (2010-03-27)
Y Han, B Sun, B Jiang, X Hu, M I Spranger, Y Zhang, Y Zhao
ABSTRACT
This study examined the transformation pathways of ginsenosides G-Rb(1) , G-Rb(3) , and G-Rc by the fungus Fusarium sacchari. Ginsenosides G-Rb(1) , G-Rb(3) and G-Rc were isolated from leaves of Radix notoginseng, and their structural identification was confirmed using NMR. Transformation of G-Rb(1) , G-Rb(3) and G-Rc by Fusarium sacchari was respectively experimented. Kinetic evolutions of G-Rb(1) , G-Rb(3) and G-Rc and their metabolites during the cell incubation were monitored by HPLC analysis. High-performance liquid chromatography (HPLC) was used for monitoring the transformation kinetics of bioactive compounds during F. sacchari metabolism. Ginsenoside C-K was transformed by F. sacchari from G-Rb(1) via G-Rd or via G-F(2) , or from G-Rb(1) via firstly Rd and then G-F(2) , and C-Mx was transformed by F. sacchari or directly from Rb(3) , or from Rb(3) via Gy-IX, while G-Mc was transformed by F. sacchari directly from G-Rc. Furthermore, C-K could be also formed from G-Rc via notoginsenoside Fe (N-Fe). The results showed an important practical application in the preparation of ginsenoside C-K. As our precious research indicated C-K possessed much more antitumor activities than C-Mx and G-Mc, so according to the transformation pathways proposed by this work, the production of antitumor compound C-K may be performed by biotransformation of G-Rb(1) previously isolated from PNLS.
Sigma-Aldrich
Ginsenoside-Rc from Panax ginseng (Korean ginseng) root, triterpenoid saponin, ≥98% (HPLC)