MilliporeSigma
  • Heart-cutting 2D-CE with on-line preconcentration for the chiral analysis of native amino acids.
Heart-cutting 2D-CE with on-line preconcentration for the chiral analysis of native amino acids.
Electrophoresis (2010-03-24)
Suzanne Anouti, Odile Vandenabeele-Trambouze, Hervé Cottet
ABSTRACT
The use of transient moving chemical reaction boundary (tMCRB) was investigated for the on-line preconcentration of native amino acids in heart-cutting 2D-CE with multiple detection points using contactless conductivity detection. The tMCRB focusing was obtained by using ammonium formate (pH 8.56) as sample matrix and acetic acid (pH 2.3) as a BGE in the first dimension of the heart-cutting 2D-CE. Different experimental parameters such as the injected volume and the concentration in ammonium formate were optimized for improving the sensitivity of detection. A stacked fraction from the first dimension was selected, isolated in the capillary, and then separated in the second dimension in the presence of a chiral selector ((+)-(18-crown-6)-2,3,11,12-tetracarboxylic acid). This on-line tMCRB preconcentration coupled with heart-cutting 2D-CE was applied with success to the chiral separation of D,L-phenylalanine, and D,L-threonine in a mixture of 22 native amino acids. The sample mixture was diluted in 0.8 M of ammonium formate, and injected at a concentration of 2.5 muM for each enantiomer with a volume corresponding to 10% of the total capillary volume. An LOD (S/N=3) of 2 muM was determined for L-threonine.