MilliporeSigma
  • Good-quality semen recovered from a paraplegic man with physostigmine salicylate treatment. A case report.
Good-quality semen recovered from a paraplegic man with physostigmine salicylate treatment. A case report.
The Journal of reproductive medicine (1991-03-01)
H Jesionowska, R Hemmings
ABSTRACT
A subcutaneous injection of physostigmine was employed to treat paraplegic anejaculation. The sperm motility was greater than in previously reported studies in which electroejaculation techniques were used for seminal emission.