MilliporeSigma
  • Discovery of a novel selective PPARgamma modulator from (-)-Cercosporamide derivatives.
Discovery of a novel selective PPARgamma modulator from (-)-Cercosporamide derivatives.
Bioorganic & medicinal chemistry letters (2010-03-12)
Akihiro Furukawa, Tsuyoshi Arita, Susumu Satoh, Kenji Wakabayashi, Shinko Hayashi, Yumi Matsui, Kazushi Araki, Masanori Kuroha, Jun Ohsumi
ABSTRACT
In an investigation of (-)-Cercosporamide derivatives with a plasma glucose-lowering effect, we found that N-benzylcarboxamide derivative 4 was a partial agonist of PPARgamma. A SAR study of the substituents on carboxamide nitrogen afforded the N-(1-naphthyl)methylcarboxamide derivative 23 as the most potent selective PPARgamma modulator. An X-ray crystallography study revealed that compound 23 bounded to the PPARgamma ligand binding domain in a unique way without any interaction with helix12. Compound 23 displayed a potent plasma glucose-lowering effect in db/db mice without the undesirable increase in body fluid and heart weight that is typically observed when PPARgamma full agonists are administrated.
Sigma-Aldrich
Cercosporamide from Cercosporidium henningsii, ≥98% (HPLC)