MilliporeSigma
  • Role of CREB in the regulatory action of sarsasapogenin on muscarinic M1 receptor density during cell aging.
Role of CREB in the regulatory action of sarsasapogenin on muscarinic M1 receptor density during cell aging.
FEBS letters (2010-03-11)
Haiyan Hu, Rui Zhang, Yongfang Zhang, Zongqin Xia, Yaer Hu
ABSTRACT
This work studied the role of cyclic AMP responsive element binding protein (CREB) in the up-regulation of M(1) muscarinic acetylcholine receptor (M(1) receptor) density by sarsasapogenin (ZMS) in CHO cells transfected with M(1) receptor gene (CHOm1 cells). During cell aging, sarsasapogenin elevated M(1) receptor density as well as CREB and phosphor-CREB (pCREB) levels. CREB peaked earliest, followed by pCREB and M(1) receptor density peaked last. When CREB synthesis was blocked by antisense oligonucleotides, the elevation effect of sarsasapogenin on M(1) receptor density was abolished. These results suggest that sarsasapogenin up-regulates M(1) receptor density in aged cells by promoting CREB production and phosphorylation. Furthermore, the results support the hypothesis that pCREB regulates M(1) receptor gene expression through heterodimer formation.
Sigma-Aldrich
Sarsasapogenin, ≥98%