MilliporeSigma
  • Fabrication of triacetylcellulose-SiO2 nanocomposites by surface modification of silica nanoparticles.
Fabrication of triacetylcellulose-SiO2 nanocomposites by surface modification of silica nanoparticles.
Langmuir : the ACS journal of surfaces and colloids (2010-02-18)
Young-Jae Kim, Shin-Woo Ha, Seung-Min Jeon, Dong Woo Yoo, Sung-Ho Chun, Byeong-Hyeok Sohn, Jin-Kyu Lee
ABSTRACT
We have successfully fabricated triacetylcellulose (TAC) polymer-silica nanocomposite films having up to 40 wt % of incorporated silica nanoparticles by deliberately designing a surface ligand that has a structure similar to that of polymer repeating units and effectively modifying the surface of silica nanoparticles through chemical bonding. Cross-sectional TEM analysis reveals no significant aggregation in all TAC-silica nanocomposite films. Thermal analysis results suggested that TAC-silica nanocomposites had higher T(g) and T(c) values as compared to pure TAC, and the increase in T(g) and T(c) was affected by the silica content. The transparency of all the nanocomposite films was over 80% in the visible range, confirming the excellent compatibility of nanoparticles with TAC. In this study, we enhance the interaction between nanoparticles and polymer matrices by modifying the surface of nanoparticles with a ligand that has a structure similar to that of polymer repeating units. It is expected that this method can be applied to other polymer systems to develop useful nanocomposites.
Product Description
Supelco
Cellulose triacetate, Selectophore