MilliporeSigma
  • Eimeria tenella: effects of diclazuril treatment on microneme genes expression in second-generation merozoites and pathological changes of caeca in parasitized chickens.
Eimeria tenella: effects of diclazuril treatment on microneme genes expression in second-generation merozoites and pathological changes of caeca in parasitized chickens.
Experimental parasitology (2010-02-09)
Bian-hua Zhou, Hong-wei Wang, Xiao-yang Wang, Li-fang Zhang, Ke-yu Zhang, Fei-qun Xue
ABSTRACT
The effects of diclazuril on mRNA expression levels of invasion-related microneme genes were examined in second-generation merozoites of Eimeria tenella (E. tenella) by quantitative real-time (QRT) PCR. Diclazruil treatment of infected chickens significantly decreased the number of second-generation merozoites by 65.13%, and resulted in downregulation of EtMIC genes: EtMIC1 by 65.63%, EtMIC2 by 64.12%, EtMIC3 by 56.82%, EtMIC4 by 73.48%, and EtMIC5 by 78.17%. SEM images of caecum tissue from uninfected chickens showed regular intestinal villus structure. In infected chickens, a distinct loss of the superficial epithelium, with a flattened mucosa and large-area necrosis and anabrosis, was evident. In diclazruil-treated chickens, a decrease in merozoite number and a visibly improved appearance of the caeca were noted. These improvements appeared to be mediated in part by downregulation of the expression of invasion-related EtMIC genes in response to diclazuril.
Product Description
Diclazuril for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Diclazuril, VETRANAL®, analytical standard