MilliporeSigma
  • Excited-state intramolecular hydrogen atom transfer of curcumin in surfactant micelles.
Excited-state intramolecular hydrogen atom transfer of curcumin in surfactant micelles.
The journal of physical chemistry. B (2010-02-09)
Ramkrishna Adhikary, Philip J Carlson, Tak W Kee, Jacob W Petrich
ABSTRACT
Femtosecond fluorescence upconversion experiments were performed on the naturally occurring medicinal pigment, curcumin, in anionic, cationic, and neutral micelles. In our studies, the micelles are composed of sodium dodecyl sulfate (SDS), dodecyl trimethyl ammonium bromide (DTAB), and triton X-100 (TX-100). We demonstrate that the excited-state kinetics of curcumin in micelles have a fast (3-8 ps) and slow (50-80 ps) component. While deuteration of curcumin has a negligible effect on the fast component, the slow component exhibits a pronounced isotope effect of approximately 1.6, indicating that micelle-captured curcumin undergoes excited-state intramolecular hydrogen atom transfer. Studies of solvation dynamics of curcumin in a 10 ps time window reveal a fast component (< or = 300 fs) followed by a 8, 6, and 3 ps component in the solvation correlation function for the TX-100, DTAB, and SDS micelles, respectively.
Sigma-Aldrich
Dodecyltrimethylammonium bromide, ≥98%
Supelco
Dodecyltrimethylammonium bromide, suitable for ion pair chromatography, LiChropur, ≥98.5% (AT)
Sigma-Aldrich
Dodecyltrimethylammonium chloride, purum, ≥98.0% anhydrous basis (AT)
Sigma-Aldrich
Dodecyltrimethylammonium bromide, BioXtra, ~99%
Sigma-Aldrich
Dodecyltrimethylammonium chloride, ≥99.0% (AT)